Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Engase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172573.2 | NP_766161.1 | 5 - 6 | 725 | 80 | |
GenBank mRNA | AK048667.1 | - | 5 - 6 | 727 | 80 | |
AK084254.1 | - | 5 - 6 | 858 | 80 | ||
AK167142.1 | - | 5 - 6 | 697 | 80 | ||
BC060640.1 | - | 4 - 5 | 549 | 80 | ||
BC120923.1 | - | 5 - 6 | 723 | 80 | ||
BC120924.1 | - | 5 - 6 | 727 | 40097903 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |