Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174852.3 | NP_777277.2 | 6 - 7 | 1459 | 69 | |
GenBank mRNA | AK089226.1 | - | 6 - 7 | 1523 | 69 | |
AK129383.1 | - | 3 - 4 | 606 | 69 | ||
AK141796.1 | - | 6 - 7 | 1363 | 69 | ||
AK155354.1 | - | 6 - 7 | 1500 | 69 | ||
AK170155.1 | - | 6 - 7 | 1547 | 69 | ||
AK170187.1 | - | 5 - 6 | 754 | 69 | ||
BC043080.1 | - | 6 - 7 | 1062 | 6400617 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |