Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113180.1 | NP_001106651.1 | 10 - 11 | 1745 | 130 | |
NM_019691.4 | NP_062665.3 | 10 - 11 | 1745 | 130 | ||
GenBank mRNA | AB022913.1 | - | 10 - 11 | 1479 | 130 | |
AB079073.1 | - | 9 - 10 | 1266 | 130 | ||
AB102777.1 | - | 9 - 10 | 1266 | 130 | ||
AK031568.1 | - | 10 - 11 | 1719 | 130 | ||
AK135034.1 | - | 10 - 11 | 1452 | 130 | ||
BC060697.1 | - | 10 - 11 | 1653 | 6409918 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |