Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53i13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024920.1 | NP_001020091.1 | 1 - 2 | 116 | 102 | |
GenBank mRNA | BC147324.1 | - | 1 - 2 | 114 | 9056369 | 102 |
BC147325.1 | - | 1 - 2 | 114 | 9056377 | 102 | |
BQ939386.1 | - | 1 - 2 | 116 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |