Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080849.3 | NP_543125.1 | 5 - 6 | 888 | 96 | |
GenBank mRNA | AF407579.1 | - | 5 - 6 | 838 | 96 | |
AK154358.1 | - | 5 - 6 | 851 | 96 | ||
BC070457.1 | - | 5 - 6 | 837 | 30603080 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |