Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139128.1 | NP_624354.1 | 6 - 7 | 862 | 87 | |
NM_139129.1 | NP_624355.1 | 5 - 6 | 742 | 87 | ||
NM_139130.2 | NP_624356.1 | 6 - 7 | 862 | 87 | ||
GenBank mRNA | AB070894.1 | - | 6 - 7 | 862 | 87 | |
AB070895.1 | - | 6 - 7 | 862 | 87 | ||
AB070896.1 | - | 5 - 6 | 742 | 87 | ||
AB288387.1 | - | 1 - 2 | 76 | 87 | ||
BC141247.1 | - | 5 - 6 | 819 | 9055859 | 87 | |
BC145575.1 | - | 6 - 7 | 940 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |