Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099688.2 | NP_001093158.1 | 1 - 2 | 92 | 82 | |
GenBank mRNA | BU937284.1 | - | 1 - 2 | 95 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |