Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028074.4 | NP_082350.3 | 1 - 2 | 234 | 97 | |
GenBank mRNA | AK021013.1 | - | 1 - 2 | 145 | 97 | |
AK049311.1 | - | 1 - 2 | 236 | 97 | ||
AK169816.1 | - | 1 - 2 | 180 | 98 | ||
AK171730.1 | - | 1 - 2 | 103 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |