Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CJ082096.1 | - | 3 - 4 | 323 | 87 | |
DV663185.1 | - | 3 - 4 | 303 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons |