Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020333.2 | NP_065066.2 | 10 - 11 | 1748 | 63 | |
GenBank mRNA | AF332063.1 | - | 10 - 11 | 1418 | 63 | |
AF332064.1 | - | 10 - 11 | 1418 | 63 | ||
AK122460.1 | - | 10 - 11 | 1482 | 63 | ||
AK147262.1 | - | 10 - 11 | 1750 | 63 | ||
BC054808.1 | - | 10 - 11 | 1414 | 6838880 | 63 | |
KJ535320.1 | - | 10 - 11 | 1439 | 63 | ||
KJ535321.1 | - | 10 - 11 | 1439 | 63 | ||
KJ535322.1 | - | 10 - 11 | 1595 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |