Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_001252573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_001252573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_001252574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_001252574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_001252575.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_001252575.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_144893.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
NM_144893.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043918.mm.1 |
TC0200005145.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043923.mm.1 |
NM_001252573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043923.mm.1 |
NM_001252573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043923.mm.1 |
NM_001252574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043923.mm.1 |
NM_001252574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043923.mm.1 |
TC0200005145.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NM_001252573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NM_001252573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NM_001252574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NM_001252574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NM_144893.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NM_144893.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NR_045544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
NR_045544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043914.mm.1 |
TC0200005145.mm.1 |
MTA TC |