Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018771.3 | NP_061241.1 | 2 - 3 | 377 | 78 | |
GenBank mRNA | AF061263.1 | - | 1 - 2 | 311 | 78 | |
AF104358.1 | - | 2 - 3 | 355 | 78 | ||
AK002492.1 | - | 2 - 3 | 358 | 78 | ||
AK086506.1 | - | 2 - 3 | 378 | 78 | ||
BC003490.1 | - | 2 - 3 | 377 | 2811749 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |