Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1gap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015046.2 | NP_001015046.1 | 14 - 15 | 1351 | 80 | |
GenBank mRNA | AK038834.1 | - | 4 - 5 | 414 | 80 | |
AK147290.1 | - | 15 - 16 | 1333 | 80 | ||
BC107388.2 | - | 5 - 6 | 557 | 40057754 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |