Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030113.2 | NP_084389.2 | 8 - 9 | 961 | 80 | |
GenBank mRNA | AF297029.1 | - | 8 - 9 | 865 | 80 | |
AF297030.1 | - | 8 - 9 | 924 | 80 | ||
AY179965.1 | - | 8 - 9 | 837 | 80 | ||
AY179966.1 | - | 8 - 9 | 837 | 80 | ||
AY179967.1 | - | 8 - 9 | 837 | 80 | ||
CN703944.1 | - | 1 - 2 | 148 | 80 | ||
CV783737.1 | - | 3 - 4 | 339 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |