Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200044089.mm.1 |
NM_001252585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044089.mm.1 |
NM_001252585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044089.mm.1 |
TC0200005155.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
NM_001252584.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
NM_001252585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
NM_001252585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
NM_001252587.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
NM_001281926.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
NM_027230.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044073.mm.1 |
TC0200005155.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044081.mm.1 |
NM_001252585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044081.mm.1 |
NM_001252585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044081.mm.1 |
NM_027230.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044081.mm.1 |
TC0200005155.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
NM_001252584.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
NM_001252585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
NM_001252585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
NM_001252587.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
NM_001281926.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
NM_027230.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044074.mm.1 |
TC0200005155.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
NM_001252584.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
NM_001252585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
NM_001252585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
NM_001252587.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
NM_001281926.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
NM_027230.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044075.mm.1 |
TC0200005155.mm.1 |
MTA TC |