Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012047.2 | NP_036177.1 | 12 - 13 | 1676 | 119 | |
GenBank mRNA | AF084259.1 | - | 12 - 13 | 1649 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |