Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172757.3 | NP_766345.3 | 8 - 9 | 1269 | 125 | |
GenBank mRNA | AK047782.1 | - | 8 - 9 | 1300 | 125 | |
AK052635.1 | - | 8 - 9 | 1296 | 125 | ||
AK134510.1 | - | 8 - 9 | 1274 | 125 | ||
AK136424.1 | - | 8 - 9 | 1279 | 125 | ||
AK140522.1 | - | 8 - 9 | 1287 | 125 | ||
AK163715.1 | - | 8 - 9 | 1256 | 125 | ||
AK165541.1 | - | 8 - 9 | 1163 | 125 | ||
BC030373.1 | - | 4 - 5 | 614 | 125 | ||
BC065060.1 | - | 8 - 9 | 1269 | 6402217 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |