Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53tg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271575.1 | NP_001258504.1 | 1 - 2 | 117 | 68 | |
GenBank mRNA | AK007294.1 | - | 1 - 2 | 118 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |