Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53tg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271575.1 | NP_001258504.1 | 2 - 3 | 190 | 95 | |
GenBank mRNA | AK007294.1 | - | 2 - 3 | 191 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |