Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039154.2 | NP_001034243.1 | 10 - 11 | 2245 | 81 | |
NM_001285913.1 | NP_001272842.1 | 10 - 11 | 2245 | 81 | ||
NM_001285914.1 | NP_001272843.1 | 10 - 11 | 2245 | 81 | ||
NM_007667.3 | NP_031693.2 | 10 - 11 | 2245 | 81 | ||
GenBank mRNA | AK044046.1 | - | 10 - 11 | 2203 | 81 | |
AK083092.1 | - | 10 - 11 | 2212 | 81 | ||
AK086711.1 | - | 10 - 11 | 2214 | 81 | ||
AK139334.1 | - | 10 - 11 | 2201 | 81 | ||
BC057581.1 | - | 10 - 11 | 2150 | 6416918 | 81 | |
BC060200.1 | - | 9 - 10 | 1968 | 81 | ||
X95600.1 | - | 10 - 11 | 1885 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |