Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159696.1 | NP_001153168.1 | 4 - 5 | 987 | 78 | |
NM_009335.2 | NP_033361.2 | 4 - 5 | 1046 | 78 | ||
GenBank mRNA | AK050973.1 | - | 4 - 5 | 964 | 78 | |
AK141211.1 | - | 4 - 5 | 948 | 78 | ||
AK145166.1 | - | 4 - 5 | 996 | 78 | ||
AK145603.1 | - | 4 - 5 | 941 | 78 | ||
BC003778.1 | - | 4 - 5 | 920 | 3592762 | 78 | |
KJ463836.1 | - | 4 - 5 | 842 | 78 | ||
X94694.1 | - | 4 - 5 | 1025 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |