Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144825.2 | NP_659074.2 | 3 - 4 | 903 | 139 | |
GenBank mRNA | AK138443.1 | - | 875 | 139 | ||
BC047271.1 | - | 2 - 3 | 250 | 139 | ||
BC151012.1 | - | 3 - 4 | 697 | 9087925 | 139 | |
BC151015.1 | - | 3 - 4 | 697 | 9087928 | 139 | |
BC151016.1 | - | 3 - 4 | 696 | 9087929 | 139 | |
CN717636.1 | - | 2 - 3 | 257 | 139 | ||
CN721819.1 | - | 1 - 2 | 54 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |