Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009866.4 | NP_033996.4 | 1 - 2 | 214 | 110 | |
GenBank mRNA | AK045023.1 | - | 1 - 2 | 183 | 110 | |
AK049652.1 | - | 1 - 2 | 180 | 110 | ||
BC046314.1 | - | 1 - 2 | 114 | 6312939 | 110 | |
CB246151.1 | - | 1 - 2 | 457 | 110 | ||
D31963.1 | - | 1 - 2 | 159 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |