Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146239.2 | NP_666351.2 | 3 - 4 | 628 | 85 | |
GenBank mRNA | AK078325.1 | - | 3 - 4 | 629 | 85 | |
AK214364.1 | - | 1 - 2 | 64 | 85 | ||
BC031778.1 | - | 175 | 4501115 | 85 | ||
BC049904.1 | - | 175 | 4501115 | 85 | ||
BC064815.1 | - | 3 - 4 | 613 | 6516981 | 85 | |
CD350146.1 | - | 2 - 3 | 258 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |