Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015810.2 | NP_056625.2 | 3 - 4 | 816 | 85 | |
NR_027785.1 | 3 - 4 | 669 | 85 | |||
GenBank mRNA | AF177202.1 | - | 3 - 4 | 721 | 85 | |
AK041591.1 | - | 3 - 4 | 788 | 85 | ||
AK042777.1 | - | 3 - 4 | 627 | 85 | ||
AK169977.1 | - | 3 - 4 | 1058 | 85 | ||
BC119131.1 | - | 3 - 4 | 745 | 8733880 | 85 | |
BC144810.1 | - | 3 - 4 | 620 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |