Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pecam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037551.1 | - | 4 - 5 | 972 | 81 | |
AK084527.1 | - | 3 - 4 | 561 | 81 | ||
BC008519.1 | - | 3 - 4 | 566 | 3592726 | 81 | |
BC085502.1 | - | 3 - 4 | 368 | 5720681 | 81 | |
L06039.1 | - | 3 - 4 | 451 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |