Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pecam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037551.1 | - | 7 - 8 | 1798 | 120 | |
AK084527.1 | - | 6 - 7 | 1388 | 120 | ||
AK169431.1 | - | 5 - 6 | 1087 | 120 | ||
BC008519.1 | - | 6 - 7 | 1392 | 3592726 | 120 | |
BC085502.1 | - | 6 - 7 | 1194 | 5720681 | 120 | |
L06039.1 | - | 6 - 7 | 1277 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |