Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164799.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164799.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_001164802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_024249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
NM_024249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039955.mm.1 |
TC1100004196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164799.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164799.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_001164802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_024249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
NM_024249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100039953.mm.1 |
TC1100004196.mm.1 |
MTA TC |