Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130886.3 | NP_570956.1 | 1 - 2 | 120 | 79 | |
GenBank mRNA | AK086176.1 | - | 2 - 3 | 369 | 79 | |
BC029102.1 | - | 1 - 2 | 120 | 3979883 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |