Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198785.1 | NP_001185714.1 | 12 - 13 | 4083 | 80 | |
NM_001198787.1 | NP_001185716.1 | 12 - 13 | 3855 | 80 | ||
NM_007377.4 | NP_031403.2 | 12 - 13 | 4366 | 80 | ||
GenBank mRNA | AB093253.2 | - | 12 - 13 | 4366 | 80 | |
AB288871.1 | - | 12 - 13 | 3985 | 80 | ||
AF011908.1 | - | 12 - 13 | 3822 | 80 | ||
AK036705.1 | - | 12 - 13 | 3857 | 80 | ||
AK147239.1 | - | 9 - 10 | 3876 | 80 | ||
AK155100.1 | - | 12 - 13 | 3857 | 80 | ||
AY236858.1 | - | 12 - 13 | 3861 | 80 | ||
AY236859.1 | - | 12 - 13 | 3888 | 80 | ||
BC042837.1 | - | 12 - 13 | 4296 | 80 | ||
BC052323.1 | - | 12 - 13 | 4297 | 80 | ||
BC080846.1 | - | 12 - 13 | 3937 | 6856191 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |