Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145896.1 | NP_001139368.1 | 10 - 11 | 2520 | 78 | |
NM_001145897.1 | NP_001139369.1 | 10 - 11 | 2464 | 78 | ||
NM_198671.2 | NP_941073.2 | 10 - 11 | 2652 | 78 | ||
GenBank mRNA | AK033263.1 | - | 10 - 11 | 2467 | 78 | |
AK042915.1 | - | 10 - 11 | 2469 | 78 | ||
AK122216.1 | - | 7 - 8 | 1802 | 78 | ||
AK154829.1 | - | 10 - 11 | 2499 | 78 | ||
BC043094.1 | - | 10 - 11 | 2652 | 6405464 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |