Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081337.1 | NP_001074806.1 | 6 - 7 | 2365 | 67 | |
GenBank mRNA | AK028440.1 | - | 6 - 7 | 1479 | 67 | |
AK122501.1 | - | 4 - 5 | 642 | 67 | ||
BC072593.1 | - | 8 - 9 | 2651 | 67 | ||
BC150682.1 | - | 6 - 7 | 2410 | 9087593 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |