Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010505.1 | - | 1 - 2 | 51 | 58 | |
AK012294.1 | - | 1 - 2 | 63 | 58 | ||
AK135451.1 | - | 1 - 2 | 41 | 58 | ||
AK145703.1 | - | 1 - 2 | 35 | 58 | ||
BC030840.1 | - | 1 - 2 | 63 | 3994523 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |