Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 15 - 16 | 3241 | 61 | |
GenBank mRNA | AK043903.1 | - | 13 - 14 | 2602 | 61 | |
AK220342.1 | - | 7 - 8 | 803 | 61 | ||
BC132334.1 | - | 11 - 12 | 2464 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |