Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800024531.mm.1 |
NM_175561.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800024531.mm.1 |
TC0800003186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800024529.mm.1 |
NM_175561.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800024529.mm.1 |
TC0800003186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800024532.mm.1 |
NM_175561.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800024532.mm.1 |
TC0800003186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800024533.mm.1 |
NM_175561.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800024533.mm.1 |
TC0800003186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800024530.mm.1 |
NM_175561.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800024530.mm.1 |
TC0800003186.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800024528.mm.1 |
NM_175561.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800024528.mm.1 |
TC0800003186.mm.1 |
MTA TC |