Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 25 - 26 | 4533 | 97 | |
GenBank mRNA | AK087907.1 | - | 10 - 11 | 1446 | 97 | |
AK220342.1 | - | 17 - 18 | 2095 | 97 | ||
BC068235.1 | - | 14 - 15 | 2600 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |