Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026922.1 | NP_081198.1 | 3 - 4 | 482 | 79 | |
GenBank mRNA | AK137687.1 | - | 3 - 4 | 483 | 79 | |
AK220351.1 | - | 1 - 2 | 67 | 79 | ||
EF688288.1 | - | 3 - 4 | 326 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Transporter Mus musculus Probe spans exons |