Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026922.1 | NP_081198.1 | 4 - 5 | 568 | 72 | |
GenBank mRNA | AK137687.1 | - | 4 - 5 | 569 | 72 | |
AK220351.1 | - | 2 - 3 | 153 | 72 | ||
EF688288.1 | - | 4 - 5 | 412 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Transporter Mus musculus Probe spans exons |