Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301211.1 | NP_001288140.1 | 8 - 9 | 1208 | 143 | |
NM_009677.6 | NP_033807.2 | 9 - 10 | 1217 | 143 | ||
GenBank mRNA | AK034198.1 | - | 6 - 7 | 446 | 143 | |
AK036387.1 | - | 9 - 10 | 1160 | 143 | ||
AK145821.1 | - | 8 - 9 | 1139 | 143 | ||
BC006691.1 | - | 8 - 9 | 884 | 143 | ||
BC052703.1 | - | 9 - 10 | 1090 | 5698075 | 143 | |
BC054535.1 | - | 9 - 10 | 1090 | 5698075 | 143 | |
X54424.1 | - | 8 - 9 | 1130 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |