Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010322.3 | NP_034452.3 | 14 - 15 | 2131 | 81 | |
GenBank mRNA | AF110769.1 | - | 14 - 15 | 2095 | 81 | |
AJ132012.1 | - | 14 - 15 | 1926 | 81 | ||
AK004364.1 | - | 5 - 6 | 578 | 81 | ||
AK004630.1 | - | 14 - 15 | 2105 | 81 | ||
AK004771.1 | - | 14 - 15 | 2111 | 81 | ||
AK010896.1 | - | 14 - 15 | 2105 | 81 | ||
AK150810.1 | - | 14 - 15 | 2089 | 81 | ||
AK166997.1 | - | 14 - 15 | 2103 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |