Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080708.2 | NP_001074177.1 | 6 - 7 | 494 | 75 | |
GenBank mRNA | AK158217.1 | - | 6 - 7 | 520 | 75 | |
BC130272.1 | - | 6 - 7 | 494 | 40143210 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |