Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080708.2 | NP_001074177.1 | 10 - 11 | 877 | 69 | |
GenBank mRNA | AK158217.1 | - | 10 - 11 | 903 | 69 | |
BC130272.1 | - | 10 - 11 | 877 | 40143210 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |