Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011032.2 | NP_035162.1 | 8 - 9 | 1300 | 82 | |
GenBank mRNA | AK141304.1 | - | 8 - 9 | 1237 | 82 | |
AK146542.1 | - | 8 - 9 | 1238 | 82 | ||
AK149605.1 | - | 8 - 9 | 1239 | 82 | ||
AK149965.1 | - | 8 - 9 | 1238 | 82 | ||
AK150002.1 | - | 8 - 9 | 1264 | 82 | ||
AK150422.1 | - | 8 - 9 | 1238 | 82 | ||
AK150592.1 | - | 8 - 9 | 1238 | 82 | ||
AK150755.1 | - | 8 - 9 | 1224 | 82 | ||
AK150805.1 | - | 8 - 9 | 1238 | 82 | ||
AK151351.1 | - | 8 - 9 | 1232 | 82 | ||
AK151546.1 | - | 8 - 9 | 1238 | 82 | ||
AK152141.1 | - | 8 - 9 | 1238 | 82 | ||
AK152217.1 | - | 8 - 9 | 1238 | 82 | ||
AK152791.1 | - | 8 - 9 | 1238 | 82 | ||
AK152834.1 | - | 8 - 9 | 1238 | 82 | ||
AK153365.1 | - | 8 - 9 | 1238 | 82 | ||
AK159606.1 | - | 8 - 9 | 1238 | 82 | ||
AK159729.1 | - | 8 - 9 | 1238 | 82 | ||
AK159965.1 | - | 8 - 9 | 1238 | 82 | ||
AK167757.1 | - | 1 - 2 | 76 | 82 | ||
AK167796.1 | - | 8 - 9 | 1238 | 82 | ||
AK168330.1 | - | 8 - 9 | 1236 | 82 | ||
AK168336.1 | - | 8 - 9 | 1220 | 82 | ||
AK168612.1 | - | 8 - 9 | 1238 | 82 | ||
AK168893.1 | - | 8 - 9 | 1238 | 82 | ||
AK169265.1 | - | 8 - 9 | 1235 | 82 | ||
AK170603.1 | - | 8 - 9 | 1235 | 82 | ||
BC008549.1 | - | 8 - 9 | 1226 | 3591404 | 82 | |
BC018581.1 | - | 1 - 2 | 86 | 82 | ||
BC093512.1 | - | 8 - 9 | 1217 | 5124376 | 82 | |
J05185.1 | - | 8 - 9 | 1300 | 82 | ||
X06453.1 | - | 8 - 9 | 1240 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |