Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002764.1 | NP_001002764.1 | 12 - 13 | 3196 | 65 | |
GenBank mRNA | AK129203.1 | - | 11 - 12 | 1815 | 65 | |
BC037179.1 | - | 10 - 11 | 1486 | 65 | ||
BC047279.1 | - | 12 - 13 | 3208 | 65 | ||
BC066040.1 | - | 12 - 13 | 3196 | 6410245 | 65 | |
BC077715.1 | - | 12 - 13 | 3159 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |