Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100029700.mm.1 |
NM_138950.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029700.mm.1 |
NM_138950.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029700.mm.1 |
TC1100003225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029701.mm.1 |
NM_138950.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029701.mm.1 |
NM_138950.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029701.mm.1 |
TC1100003225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029699.mm.1 |
NM_138950.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029699.mm.1 |
NM_138950.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029699.mm.1 |
TC1100003225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029698.mm.1 |
NM_138950.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029698.mm.1 |
NM_138950.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029698.mm.1 |
TC1100003225.mm.1 |
MTA TC |