Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198308.1 | NP_938050.1 | 15 - 16 | 2101 | 64 | |
GenBank mRNA | AK078515.1 | - | 9 - 10 | 1192 | 64 | |
AK129472.1 | - | 15 - 16 | 2205 | 64 | ||
AY223867.1 | - | 15 - 16 | 2101 | 64 | ||
BC120745.1 | - | 14 - 15 | 2031 | 40129668 | 64 | |
BC125425.1 | - | 14 - 15 | 2031 | 40129940 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |