Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177150.2 | NP_796124.1 | 9 - 10 | 1091 | 89 | |
GenBank mRNA | AK089172.1 | - | 9 - 10 | 1140 | 89 | |
AK090340.1 | - | 9 - 10 | 1092 | 89 | ||
AK143613.1 | - | 9 - 10 | 1054 | 89 | ||
AK164039.1 | - | 5 - 6 | 692 | 89 | ||
AK167376.1 | - | 9 - 10 | 1092 | 89 | ||
BC080832.1 | - | 7 - 8 | 1025 | 89 | ||
BC100321.1 | - | 4 - 5 | 1643 | 89 | ||
BC121824.1 | - | 9 - 10 | 1092 | 40107149 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |