Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081241.2 | NP_001074710.2 | 5 - 6 | 510 | 64 | |
GenBank mRNA | AK052316.1 | - | 5 - 6 | 459 | 64 | |
AK122573.1 | - | 5 - 6 | 484 | 64 | ||
AK184855.1 | - | 1 - 2 | 35 | 64 | ||
AK186816.1 | - | 1 - 2 | 35 | 64 | ||
BC079880.1 | - | 5 - 6 | 424 | 6852251 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |