Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252482.1 | NP_001239411.1 | 4 - 5 | 449 | 68 | |
NM_001252483.1 | NP_001239412.1 | 4 - 5 | 410 | 68 | ||
NM_001252484.1 | NP_001239413.1 | 4 - 5 | 340 | 68 | ||
NM_001252485.1 | NP_001239414.1 | 5 - 6 | 569 | 68 | ||
NM_213729.1 | NP_998894.1 | 4 - 5 | 478 | 68 | ||
GenBank mRNA | AK139002.1 | - | 4 - 5 | 338 | 68 | |
AY601909.1 | - | 4 - 5 | 478 | 68 | ||
AY601910.1 | - | 4 - 5 | 449 | 68 | ||
AY601911.1 | - | 4 - 5 | 410 | 68 | ||
AY601912.1 | - | 4 - 5 | 340 | 68 | ||
BC096427.1 | - | 5 - 6 | 523 | 4160694 | 68 | |
BC115572.1 | - | 4 - 5 | 309 | 40102153 | 68 | |
BC115573.1 | - | 4 - 5 | 309 | 40102154 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |