Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ednra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010332.2 | NP_034462.1 | 3 - 4 | 960 | 62 | |
GenBank mRNA | AF039892.1 | - | 2 - 3 | 255 | 62 | |
AK017486.1 | - | 3 - 4 | 955 | 62 | ||
AK042211.1 | - | 3 - 4 | 974 | 62 | ||
AK043210.1 | - | 3 - 4 | 857 | 62 | ||
AK135555.1 | - | 3 - 4 | 854 | 62 | ||
BC008277.1 | - | 3 - 4 | 944 | 2812426 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |